Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Brand: Thermo Scientific™ 97002141
Description
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
- One sample processing uses one tube strip containing five tubes.
- One tip comb with five tips is used for processing five samples at a time.
- One sample processing uses one tube strip containing five tubes.
- One tip comb with five tips is used for processing five samples at a time.
Specifications
Tubes and tip combs for 240 samples | |
KingFisher mL Combi 240 |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.