missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebro Electronic™ GFX 460 Contact Thermometer
Contact thermometer with fixed PT 1000-probe
Brand: Ebro Electronic™ 1340-5460A
Code : A0
Additional Details : Weight : 0.15000kg
Description
Thermometer, 1 channel, LEMOSA, without probeSpecifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
Single-use | |
Thermometer, sensor, calibration certificate, instruction manual | |
70 cm | |
Stainless Steel and Plastic | |
PT 1000 | |
0.6 kg | |
572°F | |
-58°F | |
-50°C to +300°C | |
±1°C | |
LCD |
54 x 22 x 110 mm without probe | |
54 mm | |
355 mm | |
Needle length 205 mm | |
GFX 460 | |
1°C | |
None | |
300°C | |
-50°C | |
Factory Calibrated | |
200 mm | |
162 mm |