Learn More
Contec™ Amplitude™ Zeta™ Cleanroom Wipes
Prepare for acid spills. The Contec™ Amplitude™ Zeta™ Specialty Cleanroom Wipes are excellent for use with acid spills or for use as a tray liner. Wipes are lot-numbered for traceability.
£245.00
Specifications
Product Type | Specialty Cleanroom Wipe |
---|---|
Material | Polypropylene |
Certifications/Compliance | ISO Class 6-8 |
For Use With (Application) | Cleanroom |
Description
Prepare for acid spills. The Contec™ Amplitude™ Zeta™ Specialty Cleanroom Wipes are excellent for use with acid spills or for use as a tray liner. Wipes are lot-numbered for traceability.
- Cellulose/polypropylene nonwoven wipes
- Thermally-laminated, quilted wipes feature polypropylene outer layers and a highly sorptive cellulose center
- Quilted pattern allows for segregation by appearance
- Excellent for acid spills
- Ideal for tray liners
- Lot-numbered for traceability
- Inner core of cellulose thermally sealed between two layers of meltblown polypropylene is sorptive
- Strong resistance to acids
- Three-ply quilted pattern allows for segregation by appearance
Order Info
Basis weight, 103g/m2.
Specifications
Specialty Cleanroom Wipe | |
Polypropylene | |
ISO Class 6-8 | |
Cleanroom |