Learn More
Contec™ Bucket Wringers and Frames
Alle ansehen Contec Produkte
Click to view available options
For Use With (Equipment):
Contec Edgeless VertiKlean MAX or QuickTask Mops, 5 gal. or 6.5 gal. Bucket
Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket
Packungsgröße:
Each
Product Type:
Heavy Duty Downpress Mop Wringer
Beschreibung
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
- Choose from downpress, flat or curved styles
- Available in polypropylene or stainless steel
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- Select wringer that complements mop
Spezifikation
Spezifikation
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Application) | Ideal for cleanrooms and controlled environments |
| For Use With (Equipment) | Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket |
| Includes | Stainless steel Flat Mop Wringer |
| Sterility | Non-sterile |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur