missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Edgeless™ Mop System
GREENER_CHOICEBIOPHARMA

Product Code. 16675514 Shop All Contec Products
Click to view available options
Head Type:
Edgeless
Color:
White
Quantity:
1/Bag, 20 Bags/Cs.
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 16675514

Brand: Contec™ ELMOP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clean in controlled environments and cleanrooms. The Edgeless™ Mop System is constructed from a continuous tube of 100% polyester knitted fabric. The mop heads help eliminate contamination from breakage and deterioration of traditional string mops.

  • Designed for wet mopping in controlled environments and cleanrooms
  • Ideal for use in sterile environments
  • Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption
  • Mop heads are free of any cellulosic contaminants
  • Textured mop heads feature ribbed, twisted strands for 60% more sorbent capacity than standard Edgeless mops
  • Textured knitted polyester tube mop heads help eliminate contamination from breakage and deterioration of traditional string mops
  • Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom
TRUSTED_SUSTAINABILITY

Specifications

Product Type Mop Head
Color Code White
Head Type Edgeless
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Limited-use
Quantity 1/Bag, 20 Bags/Cs.
For Use With (Equipment) Contec Edgeless Mopping System
Description Cleanroom Laundered Mop Head
For Use With (Application) Designed for use with aqueous solutions containing cleaning agents and disinfectants; compatible with wide range of chemicals
Color-coded No
Material Polyester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.