Learn More
Contec™ VertiKlean™ MAX™ Mop Heads
GREENER_CHOICEBIOPHARMA
Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.
£62.70 - £550.00
Specifications
Product Type Mop Head
Autoclavable Not Autoclavable
Disposable Single-use
View More Specs
Products 13
Product Code Brand Description Dimensions Sterility Price Quantity & Availability  
Product Code Brand Description Dimensions Sterility Price Quantity & Availability  
13408166
View Documents
Contec™
VKMX0200
Medium VertiKlean MAX Mop Head 32 x 11 x 2 cm (12.8 x 4.3 x 0.7 in.) Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16605564
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKMX0125
Regular VertiKlean MAX Ribbed Microfiber Mop Head 21 x 11 x 2 cm (8.4 x 4.3 x 0.7 in.) Non-sterile
£384.00
Pack of 48
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
16615564
GREENER_CHOICEBIOPHARMA
View Documents
Contec™
VKMX0175
Regular VertiKlean MAX Ribbed Microfiber Mop Head 21 x 11 x 2 cm (8.4 x 4.3 x 0.7 in.) Sterile
£550.00
Pack of 48
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15684937
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKMX0225
Medium VertiKlean MAX Ribbed Microfiber Mop Head 32 x 11 x 2 cm (12.8 x 4.3 x 0.7 in.) Non-sterile
£268.00
Pack of 28
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15585040
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKMX0275
Medium VertiKlean MAX Ribbed Microfiber Mop Head 32 x 11 x 2 cm (12.8 x 4.3 x 0.7 in.) Sterile
£482.00
Pack of 28
In Stock
to see stock.
Edge
Add to basket
15610496
GREENER_CHOICEBIOPHARMA
View Documents
Contec™
VKMX0325
Large VertiKlean MAX Ribbed Microfiber Mop Head 38 x 11 x 2 cm (15 x 4.3 x 0.7 in.) Non-sterile
£362.00
Each
Estimated Shipment: 18-06-2025
to see stock.
Add to basket
16625564
GREENER_CHOICEBIOPHARMA
View Documents
Contec™
VKMX0375
Large VertiKlean MAX Ribbed Microfiber Mop Head 38 x 11 x 2 cm (15 x 4.3 x 0.7 in.) Sterile
£427.00
Pack of 24
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
15646996
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKSE0200
38 x 13 x 1 cm (15 x 5 x 0.375 in.) Non-sterile
£435.00
Pack of 16
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15656996
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKSE0250
Medium VertiKlean MAX Ribbed Microfiber Mop Head with Ribbed Sealed Edge 38 x 13 x 1 cm (15 x 5 x 0.375 in.) Sterile
£430.00
Pack of 24
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
16348845
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKSE0100
VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge 24 x 13 x 1 cm (9.5 x 5 x 0.375 in.) Non-sterile
£397.00
Pack of 40
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
16358845
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKSE0150
VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge 24 x 13 x 1 cm (9.5 x 5 x 0.375 in.) Sterile
£500.00
Pack of 40
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
16426830
GREENER_CHOICEBIOPHARMA
View Documents
Contec™
VKSE0300
VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge 46 x 13 x 1 cm (18 x 5 x 0.375 in.) Non-sterile
£352.00
Pack of 20
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
16635564
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Contec™
VKSE0350
VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge 46 x 13 x 1 cm (18 x 5 x 0.375 in.) Sterile
£529.00
Pack of 20
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
Description
Description
  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Order Info
Frames, handles and bucket wringer system sold separately.
Certifications
Validated sterile to a Sterility Assurance Level of 10-6 per AAMI ISO11137:2006 Guidelines. Certificate of sterility is available.
Specifications
Specifications
Mop Head
Not Autoclavable
Single-use
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Contec™ VertiKlean™ MAX™ Mop Heads