missing translation for 'onlineSavingsMsg'
Learn More

Contec™ CONTEXT™ Cleanroom Paper and Notebooks
GREENER_CHOICEBIOPHARMA

CONTEXT™ Cleanroom Documentation products have been developed to provide cleanrooms with paper and notebooks that reduce the risk of contamination. Available in 8.5 x 11 in., A4, and 11 x 17 in. sizes.

Brand:  Contec™ CNBK5585W

182.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15673988

  • £310.00 / Pack of 20
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CONTEXT™ paper uses Munising Bond material made from synthetic polymer-saturated cellulose that provides optimal smoothness and natural enhanced writeability.
  • High brightness, high opacity
  • Ultra low particle generation
  • Ultra low metallic ion content
  • High tear and tensile strength
  • Heat resistant
  • Compatible with standard and high-speed laser printers
  • Compatible with wet and dry copy systems
  • Ideal for recording notes and instructions
Specifications

Specifications

Munising Bond Material from Synthetic Polyer-saturated Cellulose
5.5 in.
White
CONTEXT™ 50-Sheet Spiral College-Ruled Notebook, Plastic Cover
Autoclavable
Recording notes and work instructions
No
20 Notebooks/Cs.
18 lb.
Yes
8.5 in.
White
14 x 22 cm (5.5 x 8.5 in.)
Single-use
Standard and high-speed laser printers; wet and dry copy systems
CONTEXT™ Cleanroom Documentation
Non-sterile
67 gsm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers