missing translation for 'onlineSavingsMsg'
Learn More

SymLine Continuous Level Control Accessories

Accessories for Continuous Level Control

Brand:  SymLine 108222

35.77 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15331157

  • £44.55 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications

Specifications

Grounding cable, 1 x clamp, 1 x button plug, L = 1.5 m
1.5 m
1 x clamp, 1 x button plμg
Grounding Cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SymLine Continuous Level Control Accessories > Type: Grounding cable, 1x clamp, 1x button plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.