Learn More
SymLine Continuous Level Control Accessories
Accessories for Continuous Level Control
Brand: SymLine 108178
236.14 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.20000kg
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!Specifications
Capacitive Rod Sensor | |
For proper functionality use in combination with isolating switching amplifier 108278 & Canister 108042-S1. Housing M18 x 1, active surface material PTFE, housing material brass. Flush mountable. Sensing distance 1 to 8 mm adjustable. | |
Level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMUR |