Learn More
ICP Contract Required Detection Limits (CRDL) Standard 1, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CRDL-1
Additional Details : Weight : 0.12500kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Sb | 120μg/mL |
Co | 100μg/mL |
V | 100μg/mL |
Ni | 80μg/mL |
Cu | 50μg/mL |
Zn | 40μg/mL |
Mn | 30μg/mL |
Ag | 20μg/mL |
As | 20μg/mL |
Cr | 20μg/mL |
Tl | 20μg/mL |
Be | 10μg/mL |
Cd | 10μg/mL |
Se | 10μg/mL |
Pb | 6μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
ICP Contract Required Detection Limit Standard 1 | |
125 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5% HNO3/tr Tart. Acid/tr HF |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Components in 5% HNO3/tr. Tartaric Acid/tr. HF: Sb •120μg/mL As •20μg/mL Be •10μg/mL Cd •10μg/mL Cr •20μg/mL Co •100μg/mL Cu •50μg/mL Pb •6μg/mL Mn •30μg/mL Ni •80μg/mL Se •10μg/mL Ag •20μg/mL Tl •20μg/mL V •100μg/mL Zn •40μg/mL |
Safety and Handling
- Contract Required Detection Limits Standard
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, hydrofluoric acid
Product intended for laboratory use only