Learn More
Invitrogen™ Control BACULOSOMES™ Plus Reagent
Control BACULOSOMES™ Plus Reagent
Brand: Invitrogen™ P2315
Code : 50
Additional Details : Weight : 3.18000kg
Description
Control BACULOSOMES™ Plus Reagent is a microsomal preparation from insect cells infected with wild-type baculovirus. This product is suitable as a negative control for assays carried out using our CYP450 BACULOSOMES™ Plus Reagents.For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
BACULOSOMES™ | |
Bacterial Microsome | |
Liquid | |
Toxicology & Drug Metabolism |
Store at -80°C | |
5 mg | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.