missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Control Beads, Flow Cytometer (CE/IVD)
For use as a control for certain elements of the absolute counting process.
Brand: BD Biosciences™ 340335
Additional Details : Weight : 0.12000kg
Description
- Designed for use with in vitro diagnostic products such as BD TriTEST™ reagents and a suitably equipped flow cytometer for use as a control for certain elements of the absolute counting process.
- Specifically, a control bead value that is outside the expected range could indicate an error in pipetting or a problem with the value from the Absolute Count Beads.
- TruCOUNT Control beads are not intended as a substitute for a cellular control.
- TruCOUNT Controls can be used with the FACS Loader
Specifications
TruCOUNT Control beads are sensitive to compensation, specifically in FL2–%FL1. If lyse/no-wash procedures are not employed, care must be taken to ensure FL2 brightness is adequate. Store at 2° to 8°C. Do not use after expiration date on vial. | |
Suitable for 30 tests. | |
Flow cytometer | |
Low control bead: approx.4.72 x 104–5.25 x 104, Medium control bead: 2.351 x 105–2.635 x 105, High control bead: 9.403 x 105–1.0539 x 106 |