missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments Controlled Rate Freeze Thaw (CRFT) with Peli Case
GREENER_CHOICE

Controlled Rate Freeze Thaw (CRFT) for cryopreservation. Supplied in rigid black peli case. Interchangeable heads must be ordered seperately.

Brand:  Grant Instruments CRFT UK PACK

Product Code. 17900411

  • £24435.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Controlled rate freezer and thawer that maximizes cell survival without using liquid nitrogen.

Applications

  • Transgenic embryology research
  • Stem cell research
  • Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
  • Various mammalian cells including cardiomyocytes, adipose, liver and muscle
  • Cord blood derived stem cells
  • Adherent cells and stem cells in microplates
  • Cell suspensions in numbered/barcoded arrays
  • System integrations
  • Suitable for applications in veterinarian IVF

Key features and benefits

  • Accurate and reproducible control of cooling rates and sample temperatures
  • A focus on maximising cell survival rates
  • Controlled and customisable freezing and thawing
  • Linear and non-linear freezing profiles
  • Liquid nitrogen, alcohol and cryogen free - no need for 'topping up'
  • Interchangeable heads for flexibility (ordered separately)
  • Free of charge software allows you to design, track, export and report temperature profiles
  • Easy to use and samples can be nucleated/seeded in-situ
  • Low running costs: estimated at 1% of liquid nitrogen-controlled rate freezing and runs on a 24 V supply
  • Temperature range of -100°C to 30°C
  • Servicing and calibration options available
  • Command protocols available
Specifications

Specifications

Cryogenic Freezer/Thawer
Plastic
Plastic
Benchtop
White
2 Years
CRFT
571 x 569 x 540 mm
Internal Data Logger USB
Via Software
UPS (As Optional Extra)
3.5 A
100/240 V
170 W
Air-cooled
44.31 lb.
19.62 lb.
Interchangeable Heads
1
Aluminum
CE
No
Vials, Straws, PCR Plates
Controlled Thawing
No
507 x 510 x 498 mm
Password Protected Software
Yes
USB and Software Supplied
UK
50/60 Hz
Up to 55
Helium
20.1 kg
8.9 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers