Learn More
3M™ Cool Flow™ Fan
Optional attachment for your 3M™ 4000+ Series Reusable Half Masks. Our fan features a single button control function offering improved wearer comfort during long work shifts. It’s rechargeable and is supplied with a UK plug.
Brand: 3M™ 7100135812
503.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.14000kg Transport : UN number : 3481 Chem class : 9
Description
- Reduces heat and moisture build up
- For use with 3M™ 4000+ Series Reusable Half Masks
- Optional attachment that attaches simply and securely
- Noticeable cooling sensation on your face as you work
- Single button control function
- Rechargeable and supplied with a UK plug
Specifications
1040 Series, Standard, UK | |
Fan | |
Reusable Half Masks |