Learn More
Corning™ CoolBox™ 30 System
Provides sample cooling or freezing without ice, electricity or batteries.
Brand: Corning™ 432018
Code : NEW
Additional Details : Weight : 1.50000kg
Description
“Limited quantities remain. Buy now at a deep discount.”
(Discontinued: 432018; Suggested Replacement: 432021)
Corning CoolBox systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
- Includes: CoolBox 30 base and lid, blue cooling cartridge
- The internal cooling or freezing cores provide hours of 0.5° to 4°C cooling or sample freezing.
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
- Each CoolBox system consists of a base, cooling cartridge/core and a lid.
- To use with any 30 place Corning CoolRack Tubes Module
Specifications
Cooling: 10 hours with lid closed, 4 hours with lid open; Freezing: 6 hours with lid closed, 3 hours with lid open | |
Purple | |
1/Cs. | |
16.5 x 15.25 x 12.7 cm |
Modular Design | |
1/Pk. | |
CoolBox 30 base and lid, blue cooling cartridge | |
30 System Benchtop Cooling workstation |