missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolBox™ XT Systems

Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR
CoolBox XT System, SBS-Compatible CoolRack XT M24
Holder and One Cooling Core
Holder and Two Cooling Cores
Product Line:
CoolBox 2XT
CoolBox 2XT PCR system
CoolBox XT
CoolBox XT M24 system
CoolBox XT PCR Strip system
Unit Size:
Each
Description
Corning CoolBox 2XT systems are bench top cooling workstations that accomodate two CoolRacks and provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
- Includes two (2) XT Cooling Cores for 0.5° to 4°C cooling
- Accommodates two (2) sample modules
- Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
- To use with any 30 place Corning CoolRack Tubes Module
- The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Specifications
Specifications
| Description | Benchtop cooling system |
| Color | Orange |
| Type | Cool Box |
| For Use With (Application) | Enables Cooling for 16 Hours, Freezing for 8 Hours |
| Holds | 30 Place |
| Includes | Holder and Two Cooling Cores |
| Product Line | CoolBox 2XT |
| Weight (Metric) | 5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction