missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolBox™ XT Systems
BIOPHARMA

Product Code. 15592791
Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR
CoolBox XT System, SBS-Compatible CoolRack XT M24
Holder and One Cooling Core
Holder and Two Cooling Cores
Product Line:
CoolBox 2XT
CoolBox 2XT PCR system
CoolBox XT
CoolBox XT M24 system
CoolBox XT PCR Strip system
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15592791

Brand: Corning™ 432024

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provide sample cooling or freezing without ice, electricity, or batteries.

Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
TRUSTED_SUSTAINABILITY

Specifications

Description Benchtop cooling system
Color Pink
Type Cool Box
For Use With (Application) Enables Cooling for 16 Hours, Freezing for 8 Hours
Holds 30 Place
Includes Holder and One Cooling Core
Product Line CoolBox XT
Weight (Metric) 3 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.