Learn More
Corning™ CoolCell™ SV2 and SV10 Freezing Containers
Ensure standardized controlled-rate of -1°C/minute cell freezing in a -80°C freezer without alcohol or any fluids. Utilizes thermo-conductive alloy core and highly-insulative outer material to control rate of heat removal and provide reproducible cell cryopreservation.
Brand: Corning™
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing
Specifications
Purple | |
Freezer Box | |
1/Pk., 1/Cs. |
Round | |
12 (2 mL Serum Vials) | |
0.5 kg |