Learn More
Fisherbrand™ CoolCube Microtube and PCR Tube Cooler
Protects enzymes and other temperature-sensitive samples from degradation, while working on the laboratory bench. Microtube and PCR Tube Cooler activates overnight in freezer and provides temporary cold storage by eliminating the need for ice buckets.
Brand:  Fisherbrand™ R1002-G-FIS
Product Code. 16321338
  • £138.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for temporarily storing temperature-sensitive samples while setting up reactions
  • Maintains 0°C for 4 hours
  • Dual-sided
  • Holds up to 36 x 1.5/2.0 mL microtubes
  • Other side can hold 0.2 mL PCR tubes and strips or a 96-well PCR plate with additional capacity for 10 x 1.5/2.0 mL microtubes
Specifications
Specifications
14.3 x 14.3 x 6.4 cm (5.6 x 5.6 x 2.5 in.)
2 lb.
0°C for 4 hours
Not Autoclavable
Keeping samples cold on the bench without an ice bucket
1 kg
Light Gray
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ CoolCube Microtube and PCR Tube Cooler > Dimensions (L x W x H): 14.3 x 14.3 x 6.4 cm (5.6 x 5.6 x 2.5 in.)