missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Extension Collar for CoolBox™ 2XT System
BIOPHARMA

For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.

Brand:  Corning™ 432090

 View more versions of this product

Product Code. 15552861

  • £119.00 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Lid closure not possible with addition of extension collar

This item is not returnable. View return policy

Description

Description

For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Extension Collar
1/Pk.
Extend the height when using tall tube modules
Interior: 21.0 x 14.3 x 7.0 cm (8.2 x 5.6 x 2.7 in.)
Pink
1/Cs.
CoolBox 2XT System
26.7 x 20.0 x 7.0 cm (6.5 x 7.8 x 2.7 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.