missing translation for 'onlineSavingsMsg'
Learn More
Copper, 99.9%, Thermo Scientific Chemicals Product Code.: 10338440

Copper, 99.9%, Thermo Scientific Chemicals

Product Code. 10338440
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10338440

Brand: Thermo Scientific Acros 207691000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Assay Percent Range 99.9%
Linear Formula Cu
Quantity 100 g
Fieser 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Form Granular
Chemical Name or Material Copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466584 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog Number207690025,207691000,207695000
Lot NumberA0461659 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number207690025,207691000,207695000
Lot NumberA0459764 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number207690025,207691000,207695000
Lot NumberA0457840 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number207690025,207691000,207695000
Lot NumberA0456261 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number207690025,207691000,207695000
Product Identifier
  • Copper

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper, 99.9%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.