missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99+%, turnings, Thermo Scientific Chemicals

Catalog Number 10701171
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1083.0°C
Boiling Point 2595.0°C
Density 8.92
Assay Percent Range 99+%
Linear Formula Cu
Quantity 250 g
Fieser 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
Merck Index 15, 2504
Formula Weight 63.54
Percent Purity ≥99%
Packaging Glass bottle
Form Turnings
Chemical Name or Material Copper, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'