missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper aluminum oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Applications for copper aluminum oxide nanocrystalsare included in microbatteries, in micro-electronics, circuits and in nanowire, and in nanofluids.It is also a promising material for thermoelectric devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Formula Weight | 181.5 |
Percent Purity | 99.5% |
Assay | (metals basis) |
Chemical Name or Material | Copper aluminum oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'