missing translation for 'onlineSavingsMsg'
Learn More
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11389837

Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11389837
100 x 100 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 300 mm
This item is not returnable. View return policy

Product Code. 11389837

Brand: Thermo Scientific Alfa Aesar 042189.GH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 2.2g/50x50mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored foil
Assay Percent Range 99.999% (Metals basis)
Quantity 100 x 100 mm
Odor Odorless
Vapor Pressure 0 hPa
Form Foil
Chemical Name or Material Copper

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ19K021 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number042189.FI,042189.GH,042189.HG
Lot NumberS04K002 Certificate TypeCertificate of Analysis Date26/06/2024 Catalog Number042189.FI,042189.GH,042189.HG
Lot NumberS01I021 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number042189.FI,042189.GH,042189.HG
Lot NumberW21D045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042189.FI,042189.GH,042189.HG
Lot NumberM06B016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042189.FI,042189.GH,042189.HG

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.