missing translation for 'onlineSavingsMsg'
Learn More
Copper foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis), Oxygen free, Thermo Scientific Chemicals Product Code.: 11415170

Copper foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis), Oxygen free, Thermo Scientific Chemicals

Product Code. 11415170
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
50 x 250 mm
This item is not returnable. View return policy

Product Code. 11415170

Brand: Thermo Scientific Alfa Aesar 036395.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Weight ≈11.2g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Copper foil, 0.5mm (0.02 in.) thick, Oxygen free

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis), Oxygen free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.