missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide, 98%, Thermo Scientific Chemicals

Product Code. 11397347
Click to view available options
Quantity:
250 g
1 kg

Catalyst

It finds many applications in organic synthesis, as a catalyst and a reagent. It is used as a catalyst in Sandmeyer reaction for preparing aryl bromides from arylamines through diazonium salts. It forms complexes with Lewis bases, such as dimethylsulfide, which is a precursor to organocopper reagents. Reactions involving Atom Transfer Radical Polymerization (ATRP) and Cu-catalyzed Cross-Dehydrogenative Couplings (CDC) are catalyzed by CuBr complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name bromocopper
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C
Melting Point 492°C
Color Black-Gray
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula CuBr
UN Number UN3077
Merck Index 14,2659
Solubility Information Slightly soluble in cold water; Decomposes in hot water; Soluble in HBr,HCl,and ammonium hydroxide solution.
Formula Weight 143.45
Percent Purity 98%
Sensitivity Air Sensitive
Density 4.710 g/cm3
Chemical Name or Material Copper bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.