missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals

£30.05 - £259.53

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 99
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10005543
View Documents Promotion Details
Thermo Scientific Acros
405871000
100 g Glass bottle
£30.05
100g
Please to purchase this item. Need a web account? Register with us today!
10762392
View Documents
Thermo Scientific Acros
405875000
500 g Glass bottle
£102.51
500g
Please to purchase this item. Need a web account? Register with us today!
10248473
View Documents
Thermo Scientific Acros
405870025
2.5 kg Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-89-6
99
OXBLHERUFWYNTN-UHFFFAOYSA-M
62652
chlorocopper
ClCu
MFCD00010971
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
CHEBI:53472
Cl[Cu]

Specifications

1490.0°C
7758-89-6
Fine Crystalline Powder
ClCu
MFCD00010971
15, 2650
Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone
Cl[Cu]
99
CHEBI:53472
≥90%
4.14
Copper(I) chloride, 90+%
430.0°C
Green
90+%
CuCl
01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
OXBLHERUFWYNTN-UHFFFAOYSA-M
chlorocopper
62652
99
ACS Reagent
4.1400g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-842-9

missing translation for 'rtecsNumber' : GL6990000

missing translation for 'tsca' : TSCA

Product Identifier
  • Copper(I) chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only