Learn More
Copper(I) chloride, 99%, extra pure, purified, Thermo Scientific Chemicals
£35.20 - £460.00
Chemical Identifiers
CAS | 7758-89-6 |
---|---|
Molecular Formula | ClCu |
Molecular Weight (g/mol) | 99 |
MDL Number | MFCD00010971 |
InChI Key | OXBLHERUFWYNTN-UHFFFAOYSA-M |
Synonym | cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech |
PubChem CID | 62652 |
ChEBI | CHEBI:53472 |
IUPAC Name | chlorocopper |
SMILES | Cl[Cu] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10338200
|
Thermo Scientific Acros
208390250 |
25 g | Glass bottle |
£35.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10561841
|
Thermo Scientific Acros
208391000 |
100 g | Glass bottle |
£93.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15199223
|
Thermo Scientific Acros
208395000 |
500 g | Glass bottle |
£460.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-89-6 | |
99 | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
62652 | |
chlorocopper |
ClCu | |
MFCD00010971 | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
CHEBI:53472 | |
Cl[Cu] |
Specifications
1490.0°C | |
7758-89-6 | |
Crystalline Powder | |
ClCu | |
MFCD00010971 | |
15,265 | |
Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone | |
Cl[Cu] | |
99 | |
CHEBI:53472 | |
99% | |
4.14 | |
Copper(I) chloride, 99% |
430.0°C | |
Gray-Green | |
98.5 to 101.5% (Complexometry) | |
CuCl | |
01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97 | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
chlorocopper | |
62652 | |
99 | |
Extra Pure | |
4.1400g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-842-9
missing translation for 'rtecsNumber' : GL6990000
missing translation for 'tsca' : TSCA
- Copper(I) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only