missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) iodide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11384828
Click to view available options
Quantity:
5 g
25 g
100 g

Light, Air and Moisture Sensitive

Copper(I) iodide is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-65-4
Molecular Formula CuI
MDL Number MFCD00010978
Synonym cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Specifications

Boiling Point ca 1290°C
Melting Point 605°C
Color Brown to White
Physical Form Chunks and/or Powder
Quantity 25 g
Assay Percent Range 99.998% (Metal basis)
Solubility Information Copper(I) iodide is soluble in ammonia,thiosulfate,sodium and potassium iodide solutions. Insoluble in water and dilute acids.
Formula Weight 190.45
Odor Weak odor
Refractive Index 2.346
Sensitivity Air, light, and moisture sensitive
Appearance White to pale brown chunks/powder
Density 5.63 g/cm3
Chemical Name or Material Copper(I) iodide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Copper iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'