Learn More
Copper(I) oxide, 97%, Thermo Scientific Chemicals
£21.71 - £154.77
Chemical Identifiers
CAS | 1317-39-1 |
---|---|
Molecular Formula | Cu2O |
Molecular Weight (g/mol) | 143.09 |
MDL Number | MFCD00010974 |
Synonym | Cuprous oxide |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10328480
|
Thermo Scientific Acros
197420250 |
25 g | Glass bottle |
£21.71
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10432011
|
Thermo Scientific Acros
197425000 |
500 g | Glass bottle |
£40.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10318803
|
Thermo Scientific Acros
197420025 |
2.5 kg | Glass bottle |
£154.77
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-39-1 | |
143.09 | |
Cuprous oxide |
Cu2O | |
MFCD00010974 |
Specifications
1800.0°C | |
1317-39-1 | |
Powder | |
Cu2O | |
MFCD00010974 | |
Cuprous oxide | |
143.09 | |
97% |
1232.0°C | |
Brown-Red | |
96% min. (Complexometry) | |
Cu2O | |
15, 2654 | |
Solubility in water: insoluble. Other solubilities: soluble in ammonia or min.acids | |
143.09 | |
Copper(I) oxide, 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-270-7
missing translation for 'rtecsNumber' : GL8050000
missing translation for 'tsca' : TSCA
- Copper(I) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Copper(I) oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.