missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Oxide, Honeywell Fluka™
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Red |
Quantity | 6 x 2.5 kg |
UN Number | UN3077 |
Formula Weight | 143.09 |
Percent Purity | ≥86% (Cu basis) |
Chemical Name or Material | Copper(I) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'