missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) telluride, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11352238
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12019-52-2
Molecular Formula Cu2Te
Molecular Weight (g/mol) 254.692
MDL Number MFCD00049727
InChI Key MZEWONGNQNXVKA-UHFFFAOYSA-N
Synonym copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper
PubChem CID 6914517
IUPAC Name copper;tellurium
SMILES [Cu].[Cu].[Te]

Specifications

Color Black-Blue
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 254.68
Odor Odorless
Refractive Index 1.45
Density 7.27 g/cm3
Chemical Name or Material Copper(I) telluride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'