Learn More
Copper(I) thiocyanate, 96% min, Thermo Scientific Chemicals
£35.10 - £174.00
Chemical Identifiers
CAS | 1111-67-7 |
---|---|
Molecular Formula | CHCuNS |
Molecular Weight (g/mol) | 122.632 |
MDL Number | MFCD00010980 |
InChI Key | WEKJRGBVEBCTRF-UHFFFAOYSA-N |
Synonym | copper i thio-cyanate, thiocyanate ion copper |
PubChem CID | 16211891 |
IUPAC Name | copper;thiocyanic acid |
SMILES | C(#N)S.[Cu] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329377
|
Thermo Scientific Alfa Aesar
040220.18 |
50 g |
£35.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339377
|
Thermo Scientific Alfa Aesar
040220.30 |
250 g |
£174.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as flame retardant. It is a good inorganic pigment, and is used as the antifouling paint for protecting the underwater surfaces of ships against vegetation. Manufacturing industrial chemicals, pharmaceuticals and also used in freezing solutions, electroplating , steel picking, printing, and corrosion inhibitor against acid gases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1111-67-7 | |
122.632 | |
WEKJRGBVEBCTRF-UHFFFAOYSA-N | |
16211891 | |
C(#N)S.[Cu] |
CHCuNS | |
MFCD00010980 | |
copper i thio-cyanate, thiocyanate ion copper | |
copper;thiocyanic acid |
Specifications
1084°C | |
White | |
96% min. | |
MFCD00010980 | |
Practically insoluble in water,alcohol. Soluble in NH4OH,ether | |
C(#N)S.[Cu] | |
122.632 | |
121.63 | |
2.84 g/cm3 |
1111-67-7 | |
Powder | |
CHCuNS | |
copper i thio-cyanate, thiocyanate ion copper | |
WEKJRGBVEBCTRF-UHFFFAOYSA-N | |
copper;thiocyanic acid | |
16211891 | |
Odorless | |
Copper(I) thiocyanate |
Safety and Handling
GHS H Statement
H302-H312-H332
GHS P Statement
P273-P280h-P301+P310-P315-P420a-P501a
Warning
missing translation for 'dotInformation' : DOT Class: 9, Packing Group: III
missing translation for 'einecsNumber' : 214-183-1
missing translation for 'rtecsNumber' : GL8955000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only