Learn More
Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
£21.31 - £281.40
Chemical Identifiers
CAS | 6046-93-1 |
---|---|
Molecular Formula | C4H8CuO5 |
Molecular Weight (g/mol) | 199.65 |
MDL Number | MFCD00149570 |
InChI Key | NWFNSTOSIVLCJA-UHFFFAOYSA-L |
Synonym | copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate |
PubChem CID | 165397 |
IUPAC Name | copper;diacetate;hydrate |
SMILES | O.[Cu++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10391090
|
Thermo Scientific Acros
423591000 |
100 g | Plastic bottle |
£21.31
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10473320
|
Thermo Scientific Acros
423595000 |
500 g | Plastic bottle |
£64.92
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10199729
|
Thermo Scientific Acros
423590025 |
2.5 kg | Plastic bottle |
£281.40
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
6046-93-1 | |
199.65 | |
NWFNSTOSIVLCJA-UHFFFAOYSA-L | |
165397 | |
O.[Cu++].CC([O-])=O.CC([O-])=O |
C4H8CuO5 | |
MFCD00149570 | |
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate | |
copper;diacetate;hydrate |
Specifications
240.0°C | |
6046-93-1 | |
Solid | |
(CH3CO2)2Cu·H2O | |
01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176 | |
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate | |
NWFNSTOSIVLCJA-UHFFFAOYSA-L | |
copper;diacetate;hydrate | |
165397 | |
98.0 to 102.0% | |
Authentic | |
Copper(II) acetate monohydrate |
115.0°C | |
Blue-Green or Green | |
C4H8CuO5 | |
MFCD00149570 | |
15,2614 | |
Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether | |
O.[Cu++].CC([O-])=O.CC([O-])=O | |
199.65 | |
199.65 | |
ACS Reagent | |
1.88g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
- Copper(II) acetate monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.