missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
Boiling Point | 900.0°C |
Melting Point | 498.0°C |
Color | Black-Gray to Gray |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 99% min. (Iodometry) |
Linear Formula | CuBr2 |
Fieser | 01,161; 02,84; 05,158; 06,138; 13,112; 15,100 |
Merck Index | 15, 2619 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4 |
Show More |
Safety and Handling
Product Identifier
- Copper(II) bromide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'