missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) carbonate basic, pure, 54-56% Cu, Thermo Scientific Chemicals
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 200.0°C |
Color | Green |
Physical Form | Fine Crystalline Powder |
Quantity | 25 g |
Linear Formula | CuCO3·Cu(OH)2 |
Fieser | 01,163 |
Merck Index | 15, 2621 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acids, ammonia, practically insoluble in alcohol |
Formula Weight | 221.1 |
Percent Purity | 54 to 56% Cu |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'