missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10711101
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula Cl2Cu
Molecular Weight (g/mol) 134.45
MDL Number MFCD00010972

Specifications

Boiling Point 993.0°C
Melting Point 498.0°C
Color Brown
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (CuCl2)
Linear Formula CuCl2
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 10,106; 11,267; 13,152; 14,100
Merck Index 15, 2623
Synonym Cupric chloride
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 134.45
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Copper(II) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) chloride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.