missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) chloride dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 100°C (decomposition) |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | >99% |
Solubility Information | Soluble in water,methanol and ethanol. |
Formula Weight | 170.48 (134.45anhydrous) |
Grade | ACS Reagent |
Sensitivity | Air sensitive; Hygroscopic |
Density | 2.51 |
Chemical Name or Material | Copper(II) Chloride Dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'