missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Chloride Dihydrate, Reagent grade, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | CuCl2 · 2H2O |
UN Number | UN2802 |
Formula Weight | 170.48g/mol |
Grade | Reagent |
Packaging | Poly Bottle |
Density | Vapor Density: >1 (vs air) |
Chemical Name or Material | Copper(Ii) Chloride Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'