Learn More
Copper(II)-ethylenediamine complex, 1M solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 258550010
39.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II)-ethylenediamine complex | |
14552-35-3, 7732-18-5 | |
C4H18CuN4O2 | |
MFCD00274628 | |
Bis(ethylenediamine)copper(II)hydroxide | |
217.76 | |
Glass bottle | |
1.1000g/mL |
1M Solution in Water | |
1 L | |
Cu(H2NCH2CH2NH2)2(OH)2 | |
>110°C | |
Solubility in water: soluble | |
217.76 | |
1.1 |
Safety and Handling
- Copper(II)-ethylenediamine complex, 1M solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only