missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride dihydrate, Thermo Scientific Chemicals

Product Code. 11302178
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Flux in casting gray iron

Copper(II) fluoride can also be used to form fluorobenzene from benzene, hydrochloric acid and oxygen. Using a metal fluoride that can successfully oxidize with a methyl bond. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-88-1
Molecular Formula CuF2H4O2
Molecular Weight (g/mol) 137.57
MDL Number MFCD00010977
InChI Key CTRICEWDSISGPV-UHFFFAOYSA-L
Synonym cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride
PubChem CID 24884278
IUPAC Name copper;difluoride;dihydrate
SMILES O.O.[F-].[F-].[Cu++]

Specifications

Melting Point >130°C (decomposition)
Physical Form Powder
Quantity 50 g
UN Number UN3288
Merck Index 14,2638
Solubility Information Slightly soluble in cold water, hydrolyzes to oxyfluoride in hot water
Formula Weight 137.57 (101.54 Anhydrous)
Odor Odorless
Density 2.934 g/mL
Chemical Name or Material Copper(II) fluoride dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'