missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) gluconate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 155°C to 157°C |
Quantity | 50 g |
Merck Index | 14,2640 |
Solubility Information | Soluble in water. Insoluble in acetone,alcohol and ether. |
Formula Weight | 453.85 |
Physical Form | Powder |
Chemical Name or Material | Copper(II) gluconate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'