missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) nitrate hemi(pentahydrate), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
Specifications
Specifications
Boiling Point | 170°C (decomposition) |
Melting Point | ∼115°C |
Quantity | 1000 g |
Linear Formula | Cu(NO3)2·2.5H2O |
UN Number | UN3085 |
Merck Index | 14,2643 |
Solubility Information | Soluble in water,ammonia and ethanol |
Formula Weight | 232.59 (187.55 Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'