missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hemi(pentahydrate), 98%, Thermo Scientific Chemicals

Product Code. 11437866
Click to view available options
Quantity:
250 g
1000 g
5000 g

Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

Solubility
Soluble in water, ammonia and ethanol

Notes
Incompatible with reducing agents, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
Molecular Formula CuN2O6·2.5H2O
MDL Number MFCD00149671
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Boiling Point 170°C (decomposition)
Melting Point ∼115°C
Quantity 5000 g
Linear Formula Cu(NO3)2·2.5H2O
UN Number UN3085
Merck Index 14,2643
Solubility Information Soluble in water,ammonia and ethanol
Formula Weight 232.59 (187.55 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) nitrate hemi(pentahydrate)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.
  • P375-Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : nitirc acid, copper(2+) salt, hydrate, cuperic nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.