missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11380959
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13778-31-9
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.55
MDL Number MFCD00149669
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
Synonym copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci
PubChem CID 15303652
SMILES [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Blue
Physical Form Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in ethanol and water. Insoluble in ethyl acetate.
IUPAC Name copper(2+) dinitrate
Formula Weight 187.55 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Copper(II) nitrate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'