missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Nitrate Trihydrate, 98.0 to 103% (RT), Honeywell™

Catalog Number 15626030
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
10031-43-3
CuH6N2O9
241.599
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
This item is not returnable. View return policy
10031-43-3
CuH6N2O9
241.599
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
Melting Point >114°C
Color Blue-Green
pH 3.5 to 4.5
Physical Form Solid
Quantity 250 g
Linear Formula Cu(NO3)2 · 3H2O
UN Number UN1477
Formula Weight 241.6g/mol
Percent Purity 98.0 to 103% (RT)
Odor Odorless
Packaging Poly bottle
Density 2.05g/cm3
Chemical Name or Material Copper(II) nitrate trihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'