Learn More
Copper(II) nitrate trihydrate, 99%, for analysis, Thermo Scientific Chemicals
£12.20 - £23.50
Chemical Identifiers
CAS | 10031-43-3 |
---|---|
MDL Number | MFCD00149671 |
InChI Key | SXTLQDJHRPXDSB-UHFFFAOYSA-N |
Synonym | copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem |
PubChem CID | 9837674 |
IUPAC Name | copper;dinitrate;trihydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10592031
|
Thermo Scientific Acros
207680050 |
5 g | Glass Bottle | ||||||
10124790
|
Thermo Scientific Acros
207681000 |
100 g | Plastic bottle |
£23.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10031-43-3 | |
SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
9837674 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
MFCD00149671 | |
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
copper;dinitrate;trihydrate |
Specifications
10031-43-3 | |
Fine Powder | |
CuH6N2O9 | |
MFCD00149671 | |
15, 2633 | |
SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
copper;dinitrate;trihydrate | |
9837674 | |
98.5 to 104.0% (Iodometry) | |
Copper(II) nitrate trihydrate, p.a. |
Brown to Cream | |
99% | |
Cu(NO3)2·3H2O | |
17,366 | |
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] | |
241.599 | |
241.6 | |
Analytical |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May intensify fire; oxidizer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.
Danger
missing translation for 'rtecsNumber' : GL7875000
RUO – Research Use Only