missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxalate hemihydrate, 98%, Thermo Scientific Chemicals

Product Code. 11418306
Click to view available options
Quantity:
50 g
250 g

Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5893-66-3
Molecular Formula C4H8Cu2O10
Molecular Weight (g/mol) 343.19
MDL Number MFCD00150451
InChI Key NNZNHNMQOYUANB-UHFFFAOYSA-N
Synonym cupric oxalate, copper oxalate hemihydrate
PubChem CID 122130000
IUPAC Name copper;oxalic acid;hydrate
SMILES O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Assay Percent Range 98%
Quantity 50 g
UN Number UN3288
Merck Index 14,2645
Solubility Information Soluble in NH4OH
Formula Weight 160.57 (151.51 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Copper(II) Oxalate Hemihydrate
compliance-icons
Product Identifier
  • Copper(II) oxalate hemihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Cupric oxalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.