missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) oxide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1326.0°C |
Color | Black |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 96% min. (Iodometry) |
Linear Formula | CuO |
Merck Index | 15, 2636 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol |
IUPAC Name | copper(2+) oxidandiide |
Formula Weight | 79.54 |
Show More |
Safety and Handling
Product Identifier
- Copper(II) oxide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- H412-Harmful to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'