Learn More
Copper(II) sulfate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 200.0°C |
Color | Gray |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 97.5% min. (Iodometry) |
Linear Formula | CuSO4 |
Fieser | 01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283 |
Merck Index | 15,2643 |
Solubility Information | Solubility in water: 203g/L (20°C). |
Formula Weight | 159.6 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461978 | Certificate of Analysis | 16/04/2024 | 422870025,422871000,422875000,422870050 |
A0459773 | Certificate of Analysis | 06/02/2024 | 422870100,422870025,422871000,422875000,422870050 |
A0426454 | Certificate of Analysis | 25/08/2023 | 422870100,422870025,422871000,422875000,422870050 |
A0398464 | Certificate of Analysis | 14/12/2022 | 422870100,422870025,422871000,422875000,422870050 |
A0400980 | Certificate of Analysis | 14/12/2022 | 422870100,422870025,422871000,422875000,422870050 |
Safety and Handling


- Copper(II) sulfate
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
3Hours
18Minutes
17Seconds
45
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.